missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose Gel Loading Dye (6X, Glycerol-Based, no Xylene Cyanol FF)

Catalog No. AAJ60933AC
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60933AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60933AC Supplier Thermo Scientific Chemicals Supplier No. J60933AC

6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

Solubility
Fully misicible in water.

Notes
No Xylene cyanol FF. Contains 0.25% bromophenol blue, 30% glycerol.

Specifications

Chemical Name or Material Agarose gel loading dye
Physical Form Liquid
Name Note glycerol based, Xylene Cyanol FF free
Concentration 6X
Quantity 25 mL
Solubility Information Fully misicible in water.

RUO – Research Use Only