missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Agarose Gel Loading Dye (6X, Glycerol-Based)
$137.16 - $137.16
Specifications
| Chemical Name or Material | Agarose gel loading dye |
|---|---|
| Physical Form | Liquid |
| Name Note | glycerol based |
| Concentration | 6X |
| MDL Number | MFCD00675058 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ63869AC
|
Thermo Scientific Chemicals
J63869AC |
25 mL |
Each for $137.16
|
|
|||||
Description
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAgarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.
Solubility
Fully miscible in water.
Notes
0.25% bromophenol blue, 0.25% xylene cyanol FF and 40% glycerol
Specifications
| Agarose gel loading dye | |
| glycerol based | |
| MFCD00675058 |
| Liquid | |
| 6X | |
| Fully miscible in water. |
RUO – Research Use Only