missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose Gel Loading Dye (6X, Alkaline)

Catalog No. AAJ62157AB
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62157AB 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62157AB Supplier Thermo Scientific Chemicals Supplier No. J62157AB

Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.

Solubility
Fully miscible in water.

Notes
300mM sodium hydroxide, 6mM EDTA, 0.25% bromocresol green, 0.25% xylene cyanol FF and 18% Ficoll

Specifications

Chemical Name or Material Agarose gel loading dye
Physical Form Liquid
Name Note alkaline
Concentration 6X
Quantity 10 mL
Solubility Information Fully miscible in water.

RUO – Research Use Only