missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose, Electrophoresis Grade, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210026783
Specifications
Agarose | |
9012-36-6 | |
White | |
MFCD00081294 | |
Electrophoresis |
Powder | |
1.00g/mL at 20°C | |
250 g | |
3, 6-Anhydro-α-L-galacto-β-D-galactan, FastLane agarose |
Chemical Identifiers
9012-36-6 | |
MFCD00081294 | |
3, 6-Anhydro-α-L-galacto-β-D-galactan, FastLane agarose |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30 °C)