missing translation for 'onlineSavingsMsg'
Learn More

Agarose, Electrophoresis Grade, MP Biomedicals™

Catalog No. ICN10026725
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
50 g
250 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10026725 25 g
ICN10026705 5 g
ICN10026710 10 g
ICN10026750 50 g
ICN10026783 250 g
ICN10026790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN10026725 Supplier MP Biomedicals Inc Supplier No. 0210026725

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
Synonym 3, 6-Anhydro-α-L-galacto-β-D-galactan, FastLane agarose

Specifications

Physical Form Powder
Density 1.00g/mL at 20°C
Color White
Quantity 25 g
Grade Electrophoresis
Chemical Name or Material Agarose
Recommended Storage Store at Room Temperature (15 to 30 °C)