missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose, Electrophoresis Grade, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210026710
| Quantity | 10 g |
|---|
Chemical Identifiers
| 9012-36-6 | |
| MFCD00081294 | |
| 3, 6-Anhydro-α-L-galacto-β-D-galactan, FastLane agarose |
Specifications
| Agarose | |
| 9012-36-6 | |
| White | |
| MFCD00081294 | |
| Electrophoresis |
| Powder | |
| 1.00g/mL at 20°C | |
| 10 g | |
| 3, 6-Anhydro-α-L-galacto-β-D-galactan, FastLane agarose |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30 °C)