missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, Electrophoresis Grade

CAS: 9012-36-6

$156.40 - $699.05

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6650118
View Documents
Thermo Scientific Chemicals
J6650118
50 g N/A
Only null left
 
AAJ6650130
View Documents
Thermo Scientific Chemicals
J6650130
250 g N/A
Only null left
 
Description

Description

Standard agarose suitable for most general applications. Good gel strength and clarity. Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Standard agarose suitable for most general applications. Good gel strength and clarity. Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it.

Solubility
Soluble in water(10g/L).

Notes
Gel strength (1.5% w/v): >1120g/cm^2; Gelling temperature: 36-39°C; EEO: <0.12
Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Powder
87°C to 89°C
50 g
MFCD00081294
Electrophoresis
9012-36-6
White
Hygroscopic; Moisture sensitive
Soluble in water(10g/L).
Agarose
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only