missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, Electrophoresis Grade

Catalog No. AAJ6650130
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6650130 250 g
AAJ6650118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6650130 Supplier Thermo Scientific Chemicals Supplier No. J6650130

CAS: 9012-36-6

Standard agarose suitable for most general applications. Good gel strength and clarity. Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Standard agarose suitable for most general applications. Good gel strength and clarity. Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it.

Solubility
Soluble in water(10g/L).

Notes
Gel strength (1.5% w/v): >1120g/cm^2; Gelling temperature: 36-39°C; EEO: <0.12

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Powder
Melting Point 87°C to 89°C
Color White
Quantity 250 g
Sensitivity Hygroscopic; Moisture sensitive
Solubility Information Soluble in water(10g/L).
Grade Electrophoresis
Chemical Name or Material Agarose
EINECSNumber 232-731-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only