missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose D1-LE, molecular biology grade

CAS: 9012-36-6

$217.14 - $1924.18

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2685514
View Documents
Thermo Scientific Chemicals
H2685514
25 g
Each for $217.14
Only null left
 
AAH2685522
View Documents
Thermo Scientific Chemicals
H2685522
100 g
Each for $506.45
Only null left
 
AAH2685536
View Documents
Thermo Scientific Chemicals
H2685536
500 g
Each for $1,924.18
Only null left
 
Description

Description

Agarose D1-LE, Molecular Biology Grade is used for molecular biology applications. It acts as the medium for analytical scale electrophoretic separation in agarose gel electrophoresis. It is suitable for analytical and preparative nucleic acid electrophoresis, blotting and protein electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose D1-LE, Molecular Biology Grade is used for molecular biology applications. It acts as the medium for analytical scale electrophoretic separation in agarose gel electrophoresis. It is suitable for analytical and preparative nucleic acid electrophoresis, blotting and protein electrophoresis.

Solubility
Soluble in water.

Notes
Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

9012-36-6
Hygroscopic
Soluble in water.
Agarose D1-LE
25 g
MFCD00081294
Molecular Biology
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-731-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only