missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose D1-LE, molecular biology grade

Catalog No. AAH2685536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2685536 500 g
AAH2685514 25 g
AAH2685522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2685536 Supplier Thermo Scientific Chemicals Supplier No. H2685536

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9012-36-6

Agarose D1-LE, Molecular Biology Grade is used for molecular biology applications. It acts as the medium for analytical scale electrophoretic separation in agarose gel electrophoresis. It is suitable for analytical and preparative nucleic acid electrophoresis, blotting and protein electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose D1-LE, Molecular Biology Grade is used for molecular biology applications. It acts as the medium for analytical scale electrophoretic separation in agarose gel electrophoresis. It is suitable for analytical and preparative nucleic acid electrophoresis, blotting and protein electrophoresis.

Solubility
Soluble in water.

Notes
Moisture sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Quantity 500 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Grade Molecular Biology
Chemical Name or Material Agarose D1-LE
EINECSNumber 232-731-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only