missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™
$390.39 - $1813.00
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
BP1356-100
|
Fisher BioReagents
FLBP1356100 |
100 g | Poly Bottle |
Each for $390.39
Case of 6 Each for $2,342.34
|
|
||||
|
BP1356-500
|
Fisher BioReagents
BP1356500 |
500 g | Poly Bottle |
Each for $1,813.00
Case of 6 Each for $10,878.00
|
|
||||
Description
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade)
Chemical Identifiers
| 9012-36-6 | |
| C12H18O9 |
Specifications
| Powder/Solid | |
| Cream | |
| DNase free | |
| 34.5° to 37.5°C | |
| Poly Bottle | |
| 306.12 | |
| Agarose, Broad Separation Range for DNA/RNA (500bp to 25kb) |
| 9012-36-6 | |
| 100 g | |
| 0.15 max. | |
| 1200g/cm2min. | |
| C12H18O9 | |
| DNase- and RNase-Free |
Safety and Handling
Recommended Storage : RT