missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™

Catalog No. BP1356500
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1356500 500 g Poly Bottle
BP1356100 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP1356500 Supplier Fisher BioReagents Supplier No. BP1356 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade)

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9

Specifications

Physical Form Powder/Solid
Color Cream
Quantity 500 g
DNase DNase free
Electroendosmosis (EEO) 0.15 max.
Gelation Temperature 34.5° to 37.5°C
Gel Strength 1200g/cm2min.
Packaging Poly Bottle
Formula Weight 306.12
Grade DNase- and RNase-Free
Chemical Name or Material Agarose, Broad Separation Range for DNA/RNA (500bp to 25kb)
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT