missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™

Catalog No. BP1356100
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1356100 100 g Poly Bottle
BP1356500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP1356100 Supplier Fisher BioReagents Supplier No. FLBP1356100
  • ONLINE EXCLUSIVE PRICE
    $399.97
    $172.71 / Each

    Save $227.26

    57.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • ONLINE EXCLUSIVE PRICE
    $2,399.82
    $1,036.26 / Case of 6 Each

    Save $1,363.56

    57.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9012-36-6 | Molecular Formula: C12H18O9

Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade)

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9

Specifications

Physical Form Powder/Solid
Color Cream
Quantity 100 g
DNase DNase free
Electroendosmosis (EEO) 0.15 max.
Gelation Temperature 34.5° to 37.5°C
Gel Strength 1200g/cm2min.
Packaging Poly Bottle
Formula Weight 306.12
Grade DNase- and RNase-Free
Chemical Name or Material Agarose, Broad Separation Range for DNA/RNA (500bp to 25kb)
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT