missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, for biochemistry, pure powder (Mr = - 0.17)

CAS: 9012-36-6

$220.35 - $220.35

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC239020250
SDS View Documents
Thermo Scientific Chemicals
239020250
25 g Glass Bottle
Each for $220.35
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9012-36-6
MFCD00081294

Specifications

Powder
White
0.16 to 0.19 (-Mr)
>2000g/cm2 (1.5% gel)
Glass Bottle
Solubility in water: soluble in hot water. Other solubilities: insoluble in most common organic solvents
Agarose, Pure powder (Mr = - 0.17), For biochemistry
9012-36-6
25 g
34.5° to 37.5°C (1.5% sol.)
Authentic
MFCD00081294
Biochemical
Videos
SDS
Documents

Documents

RUO – Research Use Only