missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose, for biochemistry, pure powder (Mr = - 0.17)

Catalog No. AC239020250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC239020250 25 g Glass Bottle
AC239021000 100 g Glass Bottle
AC239025000 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC239020250 Supplier Thermo Scientific Chemicals Supplier No. 239020250

CAS: 9012-36-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Powder
Color White
Quantity 25 g
Electroendosmosis (EEO) 0.16 to 0.19 (-Mr)
Gelation Temperature 34.5° to 37.5°C (1.5% sol.)
Gel Strength >2000g/cm2 (1.5% gel)
Infrared Spectrum Authentic
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in most common organic solvents
Grade Biochemical
Chemical Name or Material Agarose, Pure powder (Mr = - 0.17), For biochemistry
Show More Show Less

RUO – Research Use Only