missing translation for 'onlineSavingsMsg'
Learn More

Agar, Powder, NF, Spectrum™ Chemical

Catalog No. p-7156995
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
25 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600335 125 g Poly Bottle
18600336 500 g Poly Bottle
18600337 2.5 kg Poly Bottle
18600338 25 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600335 Supplier Spectrum Chemical Mfg Cor Supplier No. AG110125GM
Only null left

AG110, 9002-18-0

Spectrum™ Chemical Agar, Powder, NF is used in the pharmaceutical industry for many applications such as a suspending agent, surgical lubricants and in preparations of emulsions, suspensions and capsules. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9002-18-0
Molecular Formula C14H24O9
Molecular Weight (g/mol) 336.34
InChI Key GYYDPBCUIJTIBM-UHFFFAOYNA-N
IUPAC Name 2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
SMILES COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Color Beige to Brown
Quantity 125 g
Grade NF
Chemical Name or Material Agar