Learn More
Thermo Scientific Chemicals Agar, plant cell culture tested
Used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.
$185.36 - $2734.45
Specifications
| Form | Powder |
|---|---|
| Packaging Type | Plastic Bottle |
| Quantity | 100 g |
| Sensitivity | Hygroscopic; Moisture sensitive |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2672422
|
Thermo Scientific Chemicals
H2672422 |
100 g |
Each for $185.36
|
|
|||||
|
AAH2672436
|
Thermo Scientific Chemicals
H2672436 |
500 g |
Each for $692.00
|
|
|||||
|
AAH267240E
|
Thermo Scientific Chemicals
H267240E |
2500 g |
Each for $2,734.45
|
|
|||||
Description
Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Powder | |
| Plastic Bottle | |
| 100 g | |
| Hygroscopic; Moisture sensitive |
Safety and Handling
EINECSNumber : 232-658-1
Recommended Storage : Ambient temperatures
RUO – Research Use Only