missing translation for 'onlineSavingsMsg'
Learn More

Agar, Granular, NF, Spectrum™ Chemical

AG105, 9002-18-0

Supplier:  Spectrum Chemical Mfg Cor AG10525KG

 View more versions of this product

Catalog No. 18600329


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Agar, Granular, NF is used in the pharmaceutical industry for many applications such as a suspending agent, surgical lubricants and in preparations of emulsions, suspensions and capsules. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

9002-18-0
Poly Pail
2.5 kg
COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
336.34
100%
C14H24O9
GYYDPBCUIJTIBM-UHFFFAOYNA-N
2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
NF

Chemical Identifiers

9002-18-0
336.34
2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
C14H24O9
GYYDPBCUIJTIBM-UHFFFAOYNA-N
COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions