missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agar, Granular, NF, Spectrum™ Chemical
AG105, 9002-18-0
Supplier: Spectrum Chemical Mfg Cor AG105500GM
Description
Spectrum™ Chemical Agar, Granular, NF is used in the pharmaceutical industry for many applications such as a suspending agent, surgical lubricants and in preparations of emulsions, suspensions and capsules. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
| 9002-18-0 | |
| Poly Bottle | |
| 500 g | |
| COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O | |
| 336.34 |
| 100% | |
| C14H24O9 | |
| GYYDPBCUIJTIBM-UHFFFAOYNA-N | |
| 2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol | |
| NF |
Chemical Identifiers
| 9002-18-0 | |
| 336.34 | |
| 2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol |
| C14H24O9 | |
| GYYDPBCUIJTIBM-UHFFFAOYNA-N | |
| COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O |