missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Agar, Bacteriological Grade
Specially purified for use in preparing solid culture media for microbiological and bacteriological applications.
$188.88 - $188.88
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15017880
|
MP Biomedicals™
0215017880 |
100 g |
Each for $188.88
|
|
|||||
Description
- From Red alga Rhodophyceae
- Bacteriological Grade
- White to light yellow
- 100 mesh powder
- CAS: 9002-18-0
- EC No. 232-658-1