missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Agar, Bacteriological Grade
Description
- From Red alga Rhodophyceae
- Bacteriological Grade
- White to light yellow
- 100 mesh powder
- CAS: 9002-18-0
- EC No. 232-658-1
Specifications
Specifications
| Color | Off-white |
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Grade | Bacteriological |
| Product Type | Agar |
| For Use With (Application) | Used for preparing solid culture media for microbiological and bacteriological application |
| Quantity | 100 g |
| Species | Red Alga Rhodophyceae |