Learn More
Selleck Chemical Llc Enasidenib (AG-221) 5mg 1446502-11-9 DFS Item
Supplier:  Selleck Chemicals Co Ltd S82055MG
Enasidenib (AG-221) is a first-in-class, oral, potent, reversible, selective inhibitor of the IDH2 mutant enzyme. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
Catalog No. NC1173854
Only null left
Explore available promotions