Learn More
Aescin, 98%
CAS: 6805-41-0 | C55H86O24 | 1131.269 g/mol
Supplier: Thermo Scientific Chemicals J6696806
Description
It is used and studied as a vasoprotective anti-inflammatory, anti-edematous and anti-nociceptive agent.1Â It may be used to study its pharmacokinetics, safety and efficacy as a chemosensitizer and modulator of NF-κ B cell signaling
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used and studied as a vasoprotective anti-inflammatory, anti-edematous and anti-nociceptive agent.1 It may be used to study its pharmacokinetics, safety and efficacy as a chemosensitizer and modulator of NF-κ B cell signaling
Solubility
Soluble in methanol.
Notes
Desiccate at room temperature. Incompatible with oxidizing agents.
Specifications
| Aescin | |
| White | |
| MFCD00076054 | |
| 14,3696 | |
| Soluble in DMSO. | |
| CC=C(C)C(=O)OC1C(C2(C(CC1(C)C)C3=CCC4C5(CCC(C(C5CCC4(C3(CC2O)C)C)(C)CO)OC6C(C(C(C(O6)C(=O)O)OC7C(C(C(C(O7)CO)O)O)O)O)OC8C(C(C(C(O8)CO)O)O)O)C)CO)OC(=O)C | |
| 1131.269 | |
| 1131.26 | |
| Powder |
| 6805-41-0 | |
| C55H86O24 | |
| 5 g | |
| aescin | |
| AXNVHPCVMSNXNP-XJVYZZIXSA-N | |
| (2S,3S,4S,5R,6R)-6-[[(3S,4S,6aR,6bS,8S,8aR,9R,10R,14bR)-9-acetyloxy-8-hydroxy-4,8a-bis(hydroxymethyl)-4,6a,6b,11,11,14b-hexamethyl-10-[(Z)-2-methylbut-2-enoyl]oxy-1,2,3,4a,5,6,7,8,9,10,12,12a,14,14a-tetradecahydropicen-3-yl]oxy]-4-hydroxy-3-[(2R,3S,4R,5R, | |
| 133640184 | |
| 98% |
Chemical Identifiers
| 6805-41-0 | |
| 1131.269 | |
| AXNVHPCVMSNXNP-XJVYZZIXSA-N | |
| 133640184 | |
| CC=C(C)C(=O)OC1C(C2(C(CC1(C)C)C3=CCC4C5(CCC(C(C5CCC4(C3(CC2O)C)C)(C)CO)OC6C(C(C(C(O6)C(=O)O)OC7C(C(C(C(O7)CO)O)O)O)O)OC8C(C(C(C(O8)CO)O)O)O)C)CO)OC(=O)C |
| C55H86O24 | |
| MFCD00076054 | |
| aescin | |
| (2S,3S,4S,5R,6R)-6-[[(3S,4S,6aR,6bS,8S,8aR,9R,10R,14bR)-9-acetyloxy-8-hydroxy-4,8a-bis(hydroxymethyl)-4,6a,6b,11,11,14b-hexamethyl-10-[(Z)-2-methylbut-2-enoyl]oxy-1,2,3,4a,5,6,7,8,9,10,12,12a,14,14a-tetradecahydropicen-3-yl]oxy]-4-hydroxy-3-[(2R,3S,4R,5R, |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
H302+H332-H319-H335
EINECSNumber : 229-880-6
RTECSNumber : KF6296000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only