missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Aerosolv™ 360 System
Description
Includes
Premium puncturing unit, high-efficiency filter, wire, and goggles.
Requires
55 Gallon Drum
- Enclosed system—dual seal chamber eliminates the potential for content spray back, prevents VOC release and reduces gunk build up
- High-efficiency combination coalescing carbon filter captures odors, and prevents potentially harmful VOCs from being released into the air—extended filter life reduces change outs to save time and money
- Alignment cone and two-way pressure controlled, non-sparking vent pin offers the most efficient point of puncture—greatly reduces cleanup and minimizes maintenance
- Intuitive, ergonomic activation wheel design is easy to use, and with no lever handle extending over the side of the drum, removes the chance for snagged clothing or tipped drums
- Ideal for high volume applications - safely converts spent aerosol cans from solid hazardous waste to a non-hazardous, recyclable material.
- Threading directly into a standard 2-in (51-mm) bung on a 30- (110-liter) or 55-gallon (200-liter) drum, the housing accommodates a wide variety of the most common sized cans, including domed mini, standard, and jumbo.
- Sleek design with no protruding lever handle—mechanical activation wheel with rack and pinion gear increases torque making can processing easy and fast.
- Integrated lock out system prevents unauthorized use and tamper-proof retention plate secures the unit to the drum.
- Non-sparking puncture pin pierces the can and directs the liquid flow straight into the drum for a quick, controlled evacuation.
- Non-stick coating on housing minimizes static transfer to the unit while at the same time reduces clean up.
- Attach the two-stage high-efficiency combination filter to the ¾-in (19-mm) bung to capture VOCs from any remaining propellants.
- Replaceable, proprietary polymer coalescer in filter base captures the residual liquid in the gas before it reaches the activated carbon, helping extend the life of the filter.
- The top activated carbon filter cartridge features adsorption indicator which reacts to the adsorption of VOCs to indicate when the filter cartridge is ready for replacement.
- Built-in counter tracks system use for maintenance.
- Includes safety goggles and an anti-static ground wire with alligator clip.
- One 55-gallon (200-liter) drum holds approximately 4,000 spent aerosol cans.
- Punctured, discharged, cans do not meet the definition of hazardous waste per EPA 40 CFR 261.23(a)(6), and thus can be treated as recyclable scrap metal.
Compliance
EPA Compliant
Warranty
1 year
Specifications
Specifications
| Certifications/Compliance | EPA compliant |
| Capacity (English) | 55 gal. |
| Color | Black, Green |
| Quantity | 1/Pk. |
| Autoclavable | Not Autoclavable |
| Includes | Premium puncturing unit, high-efficiency filter, wire, and goggles. |
| For Use With (Application) | Recycling aerosol cans |
| For Use With (Equipment) | 55 gallon drum |
| Height (English) | 17 in. |
| Material | Aluminum |
| Show More |