missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Aerosol-Barrier Tips

Prevent cross-contamination by using superior aerosol-barrier tips.
$274.25 - $349.00
Specifications
| Filtered | Filtered |
|---|---|
| Color | Natural |
| Clean Claims | DNase, RNase, DNA, and Pyrogen Free |
| For Use With (Equipment) | Universal Fit |
| Catalog Number | Volume (Metric) | Format | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Volume (Metric) | Format | Price | Quantity & Availability | |||||
02707152
![]() |
0.1 to 5 μL | Rack |
Pack of 960 Each for $326.07
Case of 5 Pack for $1,630.35
|
|
|||||
|
02707153
|
10 μL | Bulk |
Pack of 1000 Each for $274.25
Case of 5 Pack for $1,371.25
|
|
|||||
0270733
![]() |
30 μL | Hinged Rack |
Pack of 960 Each for $349.00
Case of 5 Pack for $1,745.00
|
|
|||||
Description
Fisherbrand™ Aerosol-Barrier tips can be used universally and protects the intergrity of your samples.
- Unique small micron filter prevents aerosol contamination with no obstruction to airflow
- Use aerosol barrier tips to ensure result integrity
- Certified by independent laboratory RNase, DNase, DNA, and pyrogen free
- Natural color
Packaging Code -- BP: Bulk Pack; RI: Rack Inserts; HR: Hinged Rack; HRS: Hinged Rack, Sterile; HR/T: Hinged Rack, Trace Metal-free; LR: Lift-off Rack; LRS: Lift-off Rack, Sterile; SR: Stack Rack; IWS: Individually Wrapped, Sterile.
Specifications
| Filtered | |
| Natural | |
| DNase, RNase, DNA, and Pyrogen Free | |
| Universal Fit |