missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Aerosol-Barrier Tips
GREENER_CHOICE

Catalog No. 0270777
Change view
Click to view available options
Volume (Metric):
1 to 5 mL
200 μL (Extended)
Format:
Bulk
Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Format
0270777 200 μL (Extended) Rack
02707168 1 to 5 mL Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0270777
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prevent cross-contamination by using superior aerosol-barrier tips.

Fisherbrand™ Aerosol-Barrier tips can be used universally and protects the intergrity of your samples.

  • Unique small micron filter prevents aerosol contamination with no obstruction to airflow
  • Use aerosol barrier tips to ensure result integrity
  • RNase, DNase, DNA, and pyrogen free
  • Natural color

Packaging Code -- BP: Bulk Pack; RI: Rack Inserts; HR: Hinged Rack; HRS: Hinged Rack, Sterile; HR/T: Hinged Rack, Trace Metal-free; LR: Lift-off Rack; LRS: Lift-off Rack, Sterile; SR: Stack Rack; IWS: Individually Wrapped, Sterile.

Specifications

Volume (Metric) 200 μL (Extended)
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Autoclavable
Length (English) 3.5 in.
Length (Metric) 91 mm
Format Rack
Tip Style Extended Length
For Use With (Equipment) Universal Fit
Graduated No
Show More Show Less