Learn More
Vikan™ Aero Tote Tub Lid
Description
- Manufactured from high-density polyethylene
- Suitable for transporting, freezing, and storing
- This model has an angled edge that fits securely on angled tubs
- These lids meet all USDA specifications and are commonly used for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications
Specifications
Specifications
| Product Type | Aero Tub Lid |
| Color | White |
| Material | Polyethylene |
| Certifications/Compliance | FDA Compliant |
| Quantity | 1 |
| Dimensions (L x W x H) | 41.2 x 37.2 x 2.75 in. |