missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals AEBSF hydrochloride, 98%

Catalog No. AC328110010
Change view
Click to view available options
Quantity:
50 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC328110010 1 g Glass bottle
AC328110500 50 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC328110010 Supplier Thermo Scientific Chemicals Supplier No. 328110010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30827-99-7 | C8H10FNO2S·ClH | 239.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30827-99-7
Molecular Formula C8H10FNO2S·ClH
Molecular Weight (g/mol) 239.7
MDL Number MFCD00132962
InChI Key WRDABNWSWOHGMS-UHFFFAOYSA-N
Synonym 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
PubChem CID 186136
IUPAC Name 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 175.0°C to 177.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in absolute ethanol
Formula Weight 239.7
Percent Purity 98%
Physical Form Powder
Chemical Name or Material AEBSF hydrochloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only