Learn More
Advanced Periodic Table of the Elements
Catalog No. p-2378932
Change view
Click to view available options
Product Type:
Advanced Periodic Table Notebook
Periodic Table Chart w/o Roller
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S45525 Periodic Table Chart w/o Roller
S45525B Advanced Periodic Table Notebook
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S45525 Supplier Customer Channels Group (Ccg) Supplier No. LL1015
Presents far more information than the typical periodic table.
  • Identifies elements as noble gases, nonmetals, and various metals classes
  • Indication of physical state at atmospheric pressure and temperature by the color of the element symbol
  • Separate presentation of important physical constants
  • Table indicates the color various elements give off when heated in the flame of a Bunsen burner
  • Each element displays IUPAC name, symbol, atomic weight, oxidation states, density, boiling and melting points, number of naturally occurring radioactive isotopes, and pertinent information for the most abundant radioactive isotope
Chart measures 58 x 42 in. (147 x 107cm); available with or without spring-activated roller. Notebook size charts are 11 x 8.5 in. (28 x 21.6cm).
Specifications
Product Type Periodic Table Chart w/o Roller
Length (English) 58 in.
Length (Metric) 147.32 cm
For Use With (Application) Identifies elements as noble gases, nonmetals and various Metals classes
Subject Science