missing translation for 'onlineSavingsMsg'
Learn More

Advanced Instruments Model BR2 Bilirubin Stat-Analyzer™

Catalog No. 23046770
Change view
Click to view available options
Capacity (Metric):
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
23046770 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23046770 Supplier Advanced Instruments Supplier No. BR2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Measures total and direct bilirubin from single sample

Emplys the method of Evelyn-Malloy (after Henry) to determine bilirubin levels through spectral absorbance at 454 and 540 nm.

  • Works with infant plasma or serum samples
  • Requires only 30μL of sample
  • Total bilirubin measured immediately; direct bilirubin (measured on same sample) takes just two minutes
  • Range 0 to 30mg/dL
  • Linearity 0.2mg/dL from straight line
  • Resolution 0.1mg/dL

Specifications

Sample Type Infant Plasma or Serum
Analysis Time 2 min.
Test Range 0 to 30 mg/dL
Calibration Yes
Repeatability ±0.2 mg/dL
Resolution ±0.1 mg/dL
Capacity (Metric) 30 μL
Certifications/Compliance CE
Reagents Diazo reagent
Content And Storage -40°C to +70°C
Warranty 1 Year
Depth (English) 13 in.
Depth (Metric) 33 cm
Height (English) 7 in.
Height (Metric) 18 cm
Width (English) 11 in.
Width (Metric) 28 cm
Electrical Requirements 100 to 120/200 to 240 V 50/60 Hz
Frequency 50/60 Hz
Voltage 100 to 120/200 to 240 V
Wattage 15 w
Weight (English) 9 lb.
Weight (Metric) 4 kg
Show More Show Less