missing translation for 'onlineSavingsMsg'
Learn More

Advanced Instruments Protinol™ Serum Osmometer Controls

Catalog No. 22046781
Change view
Click to view available options
For Use With (Equipment):
3320, 3300 Micro-osmometer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22046781 3320, 3300 Micro-osmometer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22046781 Supplier Advanced Instruments Supplier No. 3MA028
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Quality control check for the clinical laboratory for osmometers from any manufacturer

  • Developed from bovine serum albumin — adjusted with nonprotein material
  • Produce consistent, reliable results in the human serum range
  • Run controls daily to check and document the osmometer's performance
  • Set of three values: 240 ±5mOsm/kg H2O (abnormal low), 280 ±5mOsm/kg H2O (normal), and 320 ±5mOsm/kg H2O (abnormal high)

Specifications

Certifications/Compliance NIST, ISO 9000, CLIA, CAP
Content And Storage 2°C to 8°C
Concentration 240, 280, 320 mOsm/kg H2O
For Use With (Equipment) 3320, 3300 Micro-osmometer
Sample Type Serum
Type 3-Level Serum Control Kit
Includes 3mL 3 vials per level
Intended Use Use in evaluating performance of osmometer in clinical laboratories testing of Human Serum samples
Quantity 9 Pack