missing translation for 'onlineSavingsMsg'
Learn More

Advanced Instruments Osmometer System Fluid

Catalog No. 22046614
Change view
Click to view available options
For Use With (Equipment):
A2O Advanced Automated Osmometer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22046614 A2O Advanced Automated Osmometer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22046614 Supplier Advanced Instruments Supplier No. 200222
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For A2O™ Advanced™ Automated Osmometer

  • 500mL bottle

Specifications

Color Clear
For Use With (Equipment) A2O Advanced Automated Osmometer
Type Osmometer System Fluid
Includes Ozonated water
Intended Use To clean the pipetting system of the A2O osmometer
Quantity 500 mL
Form Liquid