missing translation for 'onlineSavingsMsg'
Learn More

Clinitrol™ 290 Reference Solution; Advanced Instruments

Catalog No. 22046701
Change view
Click to view available options
For Use With (Equipment):
2020, 3300 and 3320 Osmometers
3D3, 3250 and 3900 osmometers
Quantity:
10 x 2 mL
10 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Quantity
22046701 3D3, 3250 and 3900 osmometers 10 x 5 mL
22046749 2020, 3300 and 3320 Osmometers 10 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22046701 Supplier Advanced Instruments Inc Supplier No. 3LA029
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Critical normal human serum midpoint of 290mOsm

  • Checks performance of freezing-point osmometers in the critical area of normal serum values
  • Contains known and controlled quantities of some of the same solutes which comprise osmotically active particles of serum
  • Pipetted into a tube and run just like normal serum
  • Runs as the first and last samples in a series of tests to monitor operator technique and instrument performance
  • Does not require mixing, reconstituting or refrigerating; one-year shelf life

Specifications

Certifications/Compliance CE
Color Clear
Content And Storage 2°C to 30°C
Concentration 290 mOsm/kg H2O
For Use With (Equipment) 3D3, 3250 and 3900 osmometers
Sample Type Serum
Type Reference Solution
Includes Sodium Chloride (NaCl) and 2-Hydroxybiphenyl Sodium Salt Tetrahydrate
Intended Use Osmometer instrument calibration and quality control
Quantity 10 x 5 mL
Form Liquid
Show More Show Less