Learn More
Advanced Instruments Advanced™ Ease-Eject™ Sampler
Catalog No. 22046740
Change view
Click to view available options
Volume (Metric):
20 μL
Catalog No. Volume (Metric)
22046740 20 μL
1 options
Catalog No. 22046740 Supplier Advanced Instruments Supplier No. 3M0825
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Model 3300 and Model 3320 Micro-Osmometers Only
Features a reduced ejection force of the sample tip, making tip installation and removal easier and more comfortable to the instrument operator
20μL sampler
Specifications
Product Type Ease-Eject Sampler
For Use With (Equipment) Advanced 3320 and 3300 Micro Osmometer Models
Volume (English) 0.0006 oz.
Volume (Metric) 20 μL