Learn More
Eisco™ Advanced Direct Vision Spectroscope with Case
Catalog No. S27719
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S27719 1 Ea.
1 options
Catalog No. S27719 Supplier Eisco™ Supplier No. PH0594A
An Advanced Direct Vision Spectroscope with Case is designed for the rapid examination of spectral composition of white light
  • Adjustable slit and draw out focusing
  • 3 element prisms
  • Completed with wooden case
Specifications
Product Type Direct Vision Spectroscope, Advanced
Description Adjustable Slit
For Use With (Application) Rapid Examination of Spectral Composition of White Light
Includes Wooden Case, 3 Element Prisms
Quantity 1 Ea.