missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Adult Body Bag with 8 Handles, 16 mil, Blue

Mopec™ partners with American Professional Manufacturing™ to provide these sturdy and durable body bags.

Supplier:  Mopec™ BE801EBL4XL

 View more versions of this product

Catalog No. 22445591


Only null left

Description

Description

  • Custom-made, lockable slider for easy opening and closing
  • Made of PVC laminated polyester
  • HD nylon zipper
  • 100% sewn with nylon thread
  • Dielectrically sealed seams
  • Available with center or envelope zipper
  • 8 handles secured by heavy-duty steel rivets
  • Leak resistance
Specifications

Chemical Identifiers

Blue
16 mil
Body Bag
238.76 cm
PVC
94 in.
>300 lb.

Specifications

Envelope Zipper
PVC
4X-Large
94 x 84 in.
84 in.
>300 lb.
213.36 cm
Blue
1 Bag/Cs.
16 mil
94 in.
Body Bag
238.76 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions