missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Adrenocorticotropic Hormone (1-39), human
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Content And Storage | Store at -20°C |
| Molecular Formula | C207H308N56O58S |
| Format | Solid |
| InChI Key | IDLFZVILOHSSID-UHFFFAOYNA-N |
| SMILES | CSCCC(NC(=O)C(CO)NC(=O)C(CC1=CC=C(O)C=C1)NC(=O)C(N)CO)C(=O)NC(CCC(O)=O)C(=O)NC(CC1=CNC=N1)C(=O)NC(CC1=CC=CC=C1)C(=O)NC(CCCNC(N)=N)C(=O)NC(CC1=CNC2=CC=CC=C12)C(=O)NCC(=O)NC(CCCCN)C(=O)N1CCCC1C(=O)NC(C(C)C)C(=O)NCC(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)NC(CCCNC(N)=N)C(=O)NC(CCCNC(N)=N)C(=O)N1CCCC1C(=O)NC(C(C)C)C(=O)NC(CCCCN)C(=O)NC(C(C)C)C(=O)NC(CC1=CC=C(O)C=C1)C(=O)N1CCCC1C(=O)NC(CC(N)=O)C(=O)NCC(=O)NC(C)C(=O)NC(CCC(O)=O)C(=O)NC(CC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CO)C(=O)NC(C)C(=O)NC(CCC(O)=O)C(=O)NC(C)C(=O)NC(CC1=CC=CC=C1)C(=O)N1CCCC1C(=O)NC(CC(C)C)C(=O)NC(CCC(O)=O)C(=O)NC(CC1=CC=CC=C1)C(O)=O |
| IUPAC Name | 4-({1-[(1-{[1-({1-[({[6-amino-1-(2-{[1-({[(5-amino-1-{[5-amino-1-({1-[(1-{2-[(1-{[5-amino-1-({1-[(1-{2-[(2-carbamoyl-1-{[({1-[(3-carboxy-1-{[2-carboxy-1-({3-carboxy-1-[(1-{[1-({3-carboxy-1-[(1-{[1-(2-{[1-({3-carboxy-1-[(1-carboxy-2-phenylethyl)carbamoyl]propyl}carbamoyl)-3-methylbutyl]carbamoyl}pyrrolidin-1-yl)-1-oxo-3-phenylpropan-2-yl]carbamoyl}ethyl)carbamoyl]propyl}carbamoyl)ethyl]carbamoyl}-2-hydroxyethyl)carbamoyl]propyl}carbamoyl)ethyl]carbamoyl}propyl)carbamoyl]ethyl}carbamoyl)methyl]carbamoyl}ethyl)carbamoyl]pyrrolidin-1-yl}-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)carbamoyl]-2-methylpropyl}carbamoyl)pentyl]carbamoyl}-2-methylpropyl)carbamoyl]pyrrolidin-1-yl}-5-carbamimidamido-1-oxopentan-2-yl)carbamoyl]-4-carbamimidamidobutyl}carbamoyl)pentyl]carbamoyl}pentyl)carbamoyl]methyl}carbamoyl)-2-methylpropyl]carbamoyl}pyrrolidin-1-yl)-1-oxohexan-2-yl]carbamoyl}methyl)carbamoyl]-2-(1H-indol-3-yl)ethyl}carbamoyl)-4-carbamimidamidobutyl]carbamoyl}-2-phenylethyl)carbamoyl]-2-(1H-imidazol-4-yl)ethyl}carbamoyl)-4-(2-{2-[2-(2-amino-3-hydroxypropanamido)-3-(4-hydroxyphenyl)propanamido]-3-hydroxypropanamido}-4-(methylsulfanyl)butanamido)butanoic acid |
| Molecular Weight (g/mol) | 4541.14 |
| Quantity | 0.5 mg |
RUO – Research Use Only