Learn More
Promega ADP-Glo Kinase Assay + p38 alpha Kinase Enzyme System, 1 each
Catalog No. prv9591
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRV9591 1 Ea.
1 options
Catalog No. PRV9591 Supplier Promega Supplier No. V9591
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant full-length human p38alpha was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. p38alpha (SAPK2A) is a member of the p38 MAPK family, which are activated by various environmental stresses and proinflammatory cytokines.
  • Easily Screen and Profile p38α Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 Ea.
Content And Storage Components at different temperatures