Learn More
Promega ADP-Glo™ Kinase Assay + EIF2AK2 Kinase Enzyme System
Recombinant human EIF2AK2 (252-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EIF2AK2 (also known as double-stranded RNA-activated protein kinase) is shown to be involved in HIV/gp120-associated neurodegeneration.
Supplier:  Promega V5329
Encompass
Catalog No. PRV5329
Only null left
Description
Description
  • Easily Screen and Profile EIF2AK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
1 Ea.
Components at different temperatures
  • EIF2AK2 Kinase Enzyme System; (V5328) 1 x 10μg
  • ADP-Glo™ Kinase Assay; (V9101) 1 x 1,000 assays
Kinase Assay + Kinase Enzyme System
Product Suggestions
Product Suggestions