missing translation for 'onlineSavingsMsg'
Learn More

Promega ADP-Glo™ Kinase Assay + DNA-PK Kinase Enzyme System

Catalog No. PRV4107
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV4107 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV4107 Supplier Promega Supplier No. V4107

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Native human DNA-Dependent Protein Kinase purified from HeLa cells. Consists of a ∼460kDa catalytic subunit and a heterodimeric DNA-binding subunit (Ku) containing an 85kDa and a 70kDa peptide (1). Gene sequence can be found at accession number NM_006904.

  • Easily Screen and Profile DNA-PK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Quantity 1 Ea.
Content And Storage
  • DNA-PK Kinase Enzyme System; (V4106) 1 x 2,500 units
  • ADP-Glo™ Kinase Assay; (V9101) 1 x 1,000 assays
Components at different temperatures
Product Type Kinase Assay + Kinase Enzyme System