Learn More
Promega ADP-Glo™ Kinase Assay + CDK6/CyclinD3 Kinase Enzyme System
Recombinant full-length human CDK6 and CyclinD3 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on both proteins.
Supplier:  Promega V4511
Encompass
Catalog No. PRV4511
Only null left
Description
Description
  • Easily Screen and Profile CDK6/CyclinD3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
1 Ea.
Components at different temperatures
  • CDK6/CyclinD3 Kinase Enzyme System; (V4510) 1 x 10μg
  • ADP-Glo™ Kinase Assay; (V9101) 1 x 1,000 assays
Kinase Assay + Kinase Enzyme System
Product Suggestions
Product Suggestions