missing translation for 'onlineSavingsMsg'
Learn More

Promega ADP-Glo™ Kinase Assay + CDK2/CyclinE1 Kinase Enzyme System

Catalog No. PRV4489
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV4489 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV4489 Supplier Promega Supplier No. V4489

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant full-length human CDK2 and CyclinE1 were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins. CDK2 is a member of the Cyclin-Dependent Kinase family that is ubiquitously expressed.

  • Easily Screen and Profile CDK2/CyclinE1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Quantity 1 Ea.
Content And Storage
  • CDK2/CyclinE1 Kinase Enzyme System; (V4488) 1 x 10μg
  • ADP-Glo™ Kinase Assay; (V9101) 1 x 1,000 assays
Components at different temperatures
Product Type Kinase Assay + Kinase Enzyme System