Learn More
Promega ADP-Glo™ Kinase Assay
Catalog No. prv6930
Encompass
Change view
Click to view available options
Quantity:
400 Assays
Catalog No. Quantity
PRV6930 400 Assays
1 options
Catalog No. PRV6930 Supplier Promega Supplier No. V6930
ADP-Glo™ Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is converted into light by Ultra-Glo™ Luciferase. The luminescent signal positively correlates with kinase activity.
  • One Assay for all your Kinase Screening Needs
  • Use with virtually any kinase
  • Stable luminescent signal
  • Sensitive to low concentrations of ADP
Specifications
Quantity 400 Assays
Content And Storage -30°C to -10°C