missing translation for 'onlineSavingsMsg'
Learn More

Adjustable Casting Frame

Catalog No. S99540
Change view
Click to view available options
Material:
Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S99540 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S99540 Supplier Customer Channels Group (Ccg) Supplier No. FCF100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Restrict the flow of casting compounds.

  • All-aluminum frame expands a full 6 in. (15.2cm)
  • Width: 1.75 in. (4.4cm)
  • Thickness: 0.06 in. (1.5mm)

Specifications

Dimensions (L x W) 6 x 1.8 in. (15 x 4.4 cm)
For Use With (Application) For flow of casting compounds
Material Aluminum
Product Type Casting Frame