missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keystone Safety White Polypropylene Lab Coats, Elastic Wrists
Description
- Lab coats provide protection against dirt, grime, and non-hazardous dry particulates
- Non-woven polypropylene material is durable and low linting
- Ideal for use in food manufacturing, light industrial, and medical environments
- Full collar, elastic wrists
- 3-pocket style available
- Packaged 30 per case
- Meets ANSI 101-1996
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | Small |
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| Disposable | Single-use |
| Sterility | Non-sterile |
| No. of Pockets | None |
| Show More |