Learn More
Keystone Safety White Polypropylene Lab Coats, Elastic Wrists
Cool and comfortable polypropylene lab coats are ideal for nonhazardous areas where a low particulate barrier is needed.
Supplier:  Keystone Safety LC3WENW2X
 View more versions of this product
Catalog No. 19-055-504
Only null left
Explore available promotions
Description
Description
  • Lab coats provide protection against dirt, grime, and non-hazardous dry particulates
  • Non-woven polypropylene material is durable and low linting
  • Ideal for use in food manufacturing, light industrial, and medical environments
  • Full collar, elastic wrists
  • 3-pocket style available
  • Packaged 30 per case
  • Meets ANSI 101-1996
Specifications
Specifications
General Purpose Lab Coat
Unisex
White
Elastic
Non-sterile
Single
LC3-WE-NW
ANSI 101-1996
Not Autoclavable
2X-Large
Polypropylene
Snaps
Single-use
3
Yes
Serged
Food, Manufacturing, Light Industrial, Medical
30/Pk.
Product Suggestions
Product Suggestions