missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keystone Safety White Polypropylene Lab Coats, Elastic Wrists
Cool and comfortable polypropylene lab coats are ideal for nonhazardous areas where a low particulate barrier is needed.
Supplier: Keystone Safety LC3WENWXLWHITE
Description
- Lab coats provide protection against dirt, grime, and non-hazardous dry particulates
- Non-woven polypropylene material is durable and low linting
- Ideal for use in food manufacturing, light industrial, and medical environments
- Full collar, elastic wrists
- 3-pocket style available
- Packaged 30 per case
- Meets ANSI 101-1996
Specifications
| General Purpose Lab Coat | |
| Unisex | |
| White | |
| Elastic | |
| Non-sterile | |
| Single | |
| LC3-WE-NW | |
| ANSI 101-1996 | |
| Not Autoclavable |
| X-Large | |
| Polypropylene | |
| Snaps | |
| Single-use | |
| 3 | |
| Yes | |
| Serged | |
| Food, Manufacturing, Light Industrial, Medical | |
| 30/Pk. |