missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety Polypropylene Bouffant Caps

Catalog No. 19055261
Change view
Click to view available options
Size:
21 in.
Color:
Green
Navy
Orange
Red
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19055261 21 in. Orange
19055463 21 in. Green
19055464 - Red
19055465 21 in. Navy
19055466 - Yellow
19055467 - Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19055261 Supplier Keystone Safety Supplier No. 110NWI1021ORANGE*

Bouffant caps keep hair in place and help ensure a clean and hygienic work area. Available in a variety of sizes and colors.

  • Polypropylene
  • 100% Latex Free
  • Economical Protection
  • Manufactured in the USA

Specifications

Product Type Bouffant
Size 21 in.
Material Polypropylene
Color Orange
Disposable Single-use
Latex Free Yes
Model 110NWI
Sterility Non-sterile
Certifications/Compliance FFDCA Certified, IES-RP-CC-003.2
Quantity 1000/Cs.
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Style Traditional
Show More Show Less