Learn More
National Public Seating ADJ LABTBL30X60 CHEMRES, OAK
Supplier:  National Public Seating SLT53060C
Encompass
The NPS Signature Frame Adjustable Height Science Lab Tables have perfect chemistry with any science classroom. These paragons of endurance are built with a choice of stylistic light oak woodgrain, or striking textured black legs beneath a 1" thick Chem-Res tabletop. The work surface is of solid black, firmly secured, high-pressure laminate, resistant to water and 13 commonly-used laboratory chemicals. Robust legs are 2 3/8" diameter, and adjust smoothly in 3/4" increments from 27"-42". Legs automatically lock at your height setting to conveniently accommodate all seating requirements. Easily assemble the Signature Tables thanks to pre-drilled pilot holes. Feet are outfitted with oversized glides, or optional casters. The Signature Series meets MAS Certified Green environmental industrial standards. Optional Book Compartment models come with two storage compartments concealable with the included side-panel aprons.
Catalog No. 361047322