Learn More
Adipic dihydrazide, 97%
CAS: 1071-93-8 | C6H14N4O2 | 174.20 g/mol
Supplier: Thermo Scientific Chemicals A1511922
Description
Adipic dihydrazide is used as a formaldehyde scavenger and reacts with formaldehyde, thereby preventing the volatilizing of formaldehyde in the air. It is also employed as a paint additive and coating additive. It is also used as an intermediate. Further, it is used for cross-linking water-based emulsions and as a hardener for certain epoxy resins, which finds application in powder coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAdipic dihydrazide is used as a formaldehyde scavenger and reacts with formaldehyde, thereby preventing the volatilizing of formaldehyde in the air. It is also employed as a paint additive and coating additive. It is also used as an intermediate. Further, it is used for cross-linking water-based emulsions and as a hardener for certain epoxy resins, which finds application in powder coating.
Solubility
Soluble in water, alcohol and acetone.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications
| Adipic dihydrazide | |
| 180°C to 182°C | |
| C6H14N4O2 | |
| 100 g | |
| adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m | |
| IBVAQQYNSHJXBV-UHFFFAOYSA-N | |
| hexanedihydrazide | |
| 66117 | |
| 97% |
| 1071-93-8 | |
| Odorless | |
| MFCD00007614 | |
| 973863 | |
| Soluble in water,alcohol and acetone. | |
| NNC(=O)CCCCC(=O)NN | |
| 174.20 | |
| 174.2 |
Chemical Identifiers
| 1071-93-8 | |
| 174.20 | |
| IBVAQQYNSHJXBV-UHFFFAOYSA-N | |
| 66117 | |
| NNC(=O)CCCCC(=O)NN |
| C6H14N4O2 | |
| MFCD00007614 | |
| adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m | |
| hexanedihydrazide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P271-P272-P280g-P302+P352-P304+P340-P312-P333+P313-P363-P501c
H317-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 213-999-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only